Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014004.1 | NP_001014026.1 | 2 - 3 | 450 | 67 | |
GenBank mRNA | BC083803.1 | - | 2 - 3 | 450 | 7109120 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |