Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014004.1 | NP_001014026.1 | 4 - 5 | 740 | 76 | |
GenBank mRNA | BC083803.1 | - | 4 - 5 | 740 | 7109120 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |