Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014004.1 | NP_001014026.1 | 7 - 8 | 1221 | 95 | |
GenBank mRNA | BC083803.1 | - | 7 - 8 | 1221 | 7109120 | 95 |
FQ219637.1 | - | 2 - 3 | 283 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |