Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243872.2 | XP_006243934.1 | 18 - 19 | 2497 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |