Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766564.1 | XP_008764786.1 | 1 - 2 | 97 | 96 | |
XM_008766566.1 | XP_008764788.1 | 1 - 2 | 97 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |