Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 7 - 8 | 860 | 59 | |
XM_006243450.2 | XP_006243512.1 | 8 - 9 | 917 | 59 | ||
GenBank mRNA | BC158861.1 | - | 7 - 8 | 853 | 7932093 | 59 |
CB547976.1 | - | 3 - 4 | 281 | 59 | ||
FQ213896.1 | - | 7 - 8 | 842 | 59 | ||
M94040.1 | - | 7 - 8 | 771 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |