Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007020.2 | NP_001007021.1 | 1 - 2 | 153 | 70 | |
NM_001201353.1 | NP_001188282.1 | 1 - 2 | 153 | 70 | ||
NM_001201354.1 | NP_001188283.1 | 1 - 2 | 153 | 70 | ||
NM_001201355.1 | NP_001188284.1 | 1 - 2 | 153 | 70 | ||
XM_006243445.2 | XP_006243507.1 | 1 - 2 | 658 | 70 | ||
XM_006243446.2 | XP_006243508.1 | 1 - 2 | 658 | 70 | ||
XM_008766200.1 | XP_008764422.1 | 1 - 2 | 658 | 70 | ||
GenBank mRNA | BC082036.1 | - | 1 - 2 | 108 | 7130894 | 70 |
FM058030.1 | - | 1 - 2 | 153 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |