Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108180.1 | NP_001101650.1 | 5 - 6 | 792 | 64 | |
XM_006243657.2 | XP_006243719.1 | 5 - 6 | 792 | 64 | ||
XM_006243658.1 | XP_006243720.1 | 2 - 3 | 243 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |