Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108180.1 | NP_001101650.1 | 6 - 7 | 880 | 74 | |
XM_006243657.2 | XP_006243719.1 | 6 - 7 | 880 | 74 | ||
XM_006243658.1 | XP_006243720.1 | 3 - 4 | 331 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |