Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008328.1 | NP_001008329.1 | 2 - 3 | 297 | 98 | |
XM_006243725.2 | XP_006243787.1 | 2 - 3 | 618 | 98 | ||
XM_008766500.1 | XP_008764722.1 | 2 - 3 | 649 | 98 | ||
GenBank mRNA | BC085863.1 | - | 2 - 3 | 297 | 7190424 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |