Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011999.1 | NP_001011999.1 | 2 - 3 | 230 | 118 | |
GenBank mRNA | BC078910.1 | - | 2 - 3 | 230 | 7131534 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |