Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277434.1 | NP_001264363.1 | 4 - 5 | 959 | 76 | |
GenBank mRNA | FQ227817.1 | - | 2 - 3 | 243 | 76 | |
FQ230804.1 | - | 2 - 3 | 243 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |