Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100725.1 | NP_001094195.1 | 5 - 6 | 496 | 104 | |
GenBank mRNA | BC089995.1 | - | 4 - 5 | 387 | 104 | |
FQ226090.1 | - | 5 - 6 | 501 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |