Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zglp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242677.2 | XP_006242739.1 | 3 - 4 | 944 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |