Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106702.1 | NP_001100172.1 | 2 - 3 | 668 | 95 | |
GenBank mRNA | BC166828.1 | - | 2 - 3 | 631 | 8373153 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |