Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108125.1 | NP_001101595.1 | 2 - 3 | 296 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |