Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133530.1 | NP_598214.1 | 1 - 2 | 132 | 99 | |
GenBank mRNA | M60616.1 | - | 1 - 2 | 105 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |