Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012213.1 | NP_001012213.1 | 2 - 3 | 223 | 91 | |
XM_006253645.2 | XP_006253707.1 | 2 - 3 | 434 | 91 | ||
XM_008771502.1 | XP_008769724.1 | 2 - 3 | 195 | 91 | ||
GenBank mRNA | BC078935.1 | - | 2 - 3 | 223 | 7108166 | 91 |
FQ209748.1 | - | 227 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |