Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053835.1 | NP_446287.1 | 2 - 3 | 265 | 96 | |
XM_006253595.2 | XP_006253657.1 | 2 - 3 | 353 | 96 | ||
GenBank mRNA | FQ213983.1 | - | 2 - 3 | 355 | 96 | |
FQ229322.1 | - | 2 - 3 | 284 | 96 | ||
M15883.1 | - | 2 - 3 | 265 | 96 | ||
M19262.1 | - | 2 - 3 | 265 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |