Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037362.1 | NP_001032439.1 | 4 - 5 | 222 | 78 | |
XM_006253563.2 | XP_006253625.1 | 3 - 4 | 353 | 78 | ||
GenBank mRNA | BC107923.1 | - | 4 - 5 | 222 | 7444531 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |