Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Potea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770390.1 | XP_008768612.1 | 12 - 13 | 1517 | 69 | |
XM_008770391.1 | XP_008768613.1 | 12 - 13 | 1517 | 69 | ||
XM_008770392.1 | XP_008768614.1 | 11 - 12 | 1343 | 69 | ||
XM_008770393.1 | XP_008768615.1 | 7 - 8 | 2259 | 69 | ||
GenBank mRNA | AB095021.1 | - | 12 - 13 | 1386 | 69 | |
BC083629.1 | - | 12 - 13 | 1494 | 7134382 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |