Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Potea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770390.1 | XP_008768612.1 | 6 - 7 | 1045 | 91 | |
XM_008770391.1 | XP_008768613.1 | 6 - 7 | 1045 | 91 | ||
XM_008770392.1 | XP_008768614.1 | 5 - 6 | 871 | 91 | ||
GenBank mRNA | AB095021.1 | - | 6 - 7 | 914 | 91 | |
BC083629.1 | - | 6 - 7 | 1019 | 7134382 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |