Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033894.1 | NP_001029066.1 | 9 - 10 | 1636 | 98 | |
XM_006251094.1 | XP_006251156.1 | 8 - 9 | 1569 | 98 | ||
GenBank mRNA | BC104677.1 | - | 9 - 10 | 1636 | 7453460 | 98 |
BC104718.1 | - | 8 - 9 | 1576 | 7381652 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |