Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106481.1 | NP_001099951.1 | 3 - 4 | 758 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |