Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kynu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053902.2 | NP_446354.1 | 7 - 8 | 628 | 133 | |
XM_008761695.1 | XP_008759917.1 | 7 - 8 | 691 | 133 | ||
GenBank mRNA | BC078762.1 | - | 7 - 8 | 611 | 7128911 | 133 |
U68168.1 | - | 7 - 8 | 629 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |