Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044255.1 | NP_001037720.1 | 3 - 4 | 552 | 142 | |
XM_006250675.2 | XP_006250737.1 | 3 - 4 | 584 | 142 | ||
GenBank mRNA | BC099827.1 | - | 3 - 4 | 552 | 7934175 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |