Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270556.1 | NP_001257485.1 | 11 - 12 | 1620 | 94 | |
NM_012806.2 | NP_036938.2 | 11 - 12 | 1647 | 94 | ||
XM_008769992.1 | XP_008768214.1 | 10 - 11 | 4439 | 94 | ||
GenBank mRNA | DQ377224.1 | - | 8 - 9 | 994 | 94 | |
FQ211777.1 | - | 11 - 12 | 1619 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |