Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108353.1 | NP_001101823.1 | 1 - 2 | 90 | 67 | |
GenBank mRNA | BC168910.1 | - | 1 - 2 | 39 | 7378144 | 67 |
FQ220996.1 | - | 1 - 2 | 65 | 67 | ||
FQ221955.1 | - | 1 - 2 | 65 | 67 | ||
FQ222120.1 | - | 1 - 2 | 66 | 67 | ||
FQ222760.1 | - | 1 - 2 | 67 | 67 | ||
FQ233729.1 | - | 1 - 2 | 66 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |