Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012033.1 | NP_001012033.1 | 4 - 5 | 812 | 78 | |
XM_008770025.1 | XP_008768247.1 | 4 - 5 | 874 | 78 | ||
GenBank mRNA | BC079418.1 | - | 4 - 5 | 812 | 7113653 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |