Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 5 - 6 | 652 | 82 | |
GenBank mRNA | D17667.1 | - | 4 - 5 | 427 | 82 | |
D38082.1 | - | 4 - 5 | 509 | 82 | ||
S75359.1 | - | 5 - 6 | 652 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |