Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031339.1 | NP_112629.1 | 16 - 17 | 2756 | 62 | |
XM_006252669.1 | XP_006252731.1 | 15 - 16 | 2426 | 62 | ||
GenBank mRNA | AB019366.1 | - | 16 - 17 | 2756 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |