Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771033.1 | XP_008769255.1 | 12 - 13 | 1855 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |