Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004255.1 | NP_001004255.1 | 2 - 3 | 257 | 65 | |
XM_006253756.2 | XP_006253818.1 | 2 - 3 | 280 | 65 | ||
GenBank mRNA | BC079344.1 | - | 2 - 3 | 257 | 7107191 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |