Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031811.3 | NP_113999.2 | 5 - 6 | 653 | 122 | |
GenBank mRNA | AF069306.1 | - | 5 - 6 | 639 | 122 | |
BC059126.1 | - | 5 - 6 | 657 | 6889440 | 122 | |
FQ213939.1 | - | 5 - 6 | 665 | 122 | ||
FQ219168.1 | - | 5 - 6 | 679 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |