Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031811.3 | NP_113999.2 | 1 - 2 | 107 | 71 | |
GenBank mRNA | AF069306.1 | - | 1 - 2 | 93 | 71 | |
BC059126.1 | - | 1 - 2 | 111 | 6889440 | 71 | |
FQ213939.1 | - | 1 - 2 | 119 | 71 | ||
FQ219168.1 | - | 1 - 2 | 133 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |