Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017224.2 | NP_058920.1 | 5 - 6 | 965 | 118 | |
XM_006230978.2 | XP_006231040.1 | 5 - 6 | 1220 | 118 | ||
GenBank mRNA | AB004559.1 | - | 5 - 6 | 1195 | 118 | |
AF008221.1 | - | 5 - 6 | 1173 | 118 | ||
BC104692.1 | - | 5 - 6 | 965 | 7109188 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |