Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286936.1 | NP_001273865.1 | 5 - 6 | 594 | 80 | |
XM_006230901.2 | XP_006230963.1 | 5 - 6 | 570 | 80 | ||
XM_008760175.1 | XP_008758397.1 | 5 - 6 | 558 | 80 | ||
GenBank mRNA | DY546174.1 | - | 3 - 4 | 314 | 80 | |
FQ227385.1 | - | 5 - 6 | 630 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |