Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181386.2 | NP_852051.2 | 3 - 4 | 468 | 69 | |
GenBank mRNA | BC081693.1 | - | 3 - 4 | 468 | 7132545 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |