Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034829.1 | NP_001030001.1 | 5 - 6 | 483 | 75 | |
XM_006230975.1 | XP_006231037.1 | 6 - 7 | 660 | 75 | ||
GenBank mRNA | AJ277751.1 | - | 5 - 6 | 483 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |