Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rorb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270958.1 | NP_001257887.1 | 8 - 9 | 1742 | 93 | |
XM_002728869.3 | XP_002728915.1 | 8 - 9 | 3394 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |