Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108513.1 | NP_001101983.1 | 6 - 7 | 877 | 70 | |
XM_006230833.1 | XP_006230895.1 | 6 - 7 | 844 | 70 | ||
GenBank mRNA | BC088464.1 | - | 6 - 7 | 808 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |