Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vopp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108630.1 | NP_001102100.1 | 3 - 4 | 348 | 78 | |
XM_006236578.2 | XP_006236640.1 | 3 - 4 | 497 | 78 | ||
GenBank mRNA | BC168709.1 | - | 3 - 4 | 341 | 8363599 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |