Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729089.4 | XP_002729135.3 | 4 - 5 | 1222 | 75 | |
XM_003749311.3 | XP_003749359.1 | 4 - 5 | 1031 | 75 | ||
XM_006232554.2 | XP_006232616.2 | 5 - 6 | 1128 | 75 | ||
XM_006232555.2 | XP_006232617.1 | 4 - 5 | 3309 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |