Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022379.1 | NP_071774.1 | 4 - 5 | 560 | 116 | |
XM_006236128.2 | XP_006236190.1 | 3 - 4 | 826 | 116 | ||
GenBank mRNA | L08812.1 | - | 4 - 5 | 560 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |