Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053686.1 | NP_446138.1 | 9 - 10 | 1498 | 98 | |
XM_006236375.2 | XP_006236437.1 | 9 - 10 | 1114 | 98 | ||
GenBank mRNA | AF160798.1 | - | 9 - 10 | 1498 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |