Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Theg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013102.1 | NP_001013120.1 | 5 - 6 | 653 | 122 | |
XM_006240857.2 | XP_006240919.1 | 5 - 6 | 673 | 122 | ||
XM_006240858.2 | XP_006240920.1 | 4 - 5 | 601 | 122 | ||
GenBank mRNA | BC083921.1 | - | 5 - 6 | 653 | 7135133 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |