Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109282.1 | NP_001102752.1 | 3 - 4 | 553 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |