Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008525.1 | NP_001008525.1 | 3 - 4 | 124 | 105 | |
XM_006240972.2 | XP_006241034.1 | 4 - 5 | 371 | 105 | ||
GenBank mRNA | BC086574.1 | - | 3 - 4 | 124 | 7306868 | 105 |
FQ232492.1 | - | 3 - 4 | 133 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |