Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138901.2 | NP_620256.1 | 3 - 4 | 436 | 102 | |
XM_008767767.1 | XP_008765989.1 | 3 - 4 | 432 | 102 | ||
GenBank mRNA | BC074020.1 | - | 3 - 4 | 436 | 7104008 | 102 |
D88364.1 | - | 3 - 4 | 817 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |