Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009504.1 | NP_001009504.1 | 4 - 5 | 1291 | 122 | |
XM_006246020.2 | XP_006246082.1 | 5 - 6 | 1609 | 122 | ||
GenBank mRNA | AY823669.1 | - | 4 - 5 | 1291 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |